• USA Home
  • Search Results

  • dichloromethane84937509211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Dichloromethane

34 Products
Synonym:   DCM, Methylene chloride
Image of Dichloromethane
CAS Number:
Empirical Formula (Hill Notation):
 CH2Cl2
Molecular Weight:
 84.93
 Beilstein/REAXYS Number:
 1730800
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Absorption
  Shelf Life
  Type
  Application(s)
  Purification method
  Product Line
  Quality clarifier
  Purity Grade
  Density
  UV-Absorption Condition
  Ice Shipping
  Agency/Method
  Solubility
  Restriction
  Autoignition temp
  Refractive Index
  Suitability
  Transmittance
  Melting Point
  Color
  Potency
  UV-Absorption Values
  Additive
  Assay
Show More

Product #
Description
270997 Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer,
34856 Dichloromethane, for HPLC, ≥99.8%, contains amylene as stabilizer,
650463 Dichloromethane, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer,
D65100 Dichloromethane, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%,
32222 Dichloromethane, puriss. p.a., ACS reagent, reag. ISO, ≥99.9% (GC),
439223 Dichloromethane, for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer,
24233 Dichloromethane, puriss., meets analytical specification of Ph. Eur., NF, ≥99% (GC),
676853 Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer,
154792 Dichloromethane, ACS spectrophotometric grade, ≥99.5%, contains 50-150 ppm amylene as stabilizer,
320269 Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer,
02575 Dichloromethane, analytical standard,
494453 Dichloromethane, biotech. grade, 99.9%, contains 40-150 ppm amylene as stabilizer,
48538 Dichloromethane, analytical standard, stabilized,
270563 Dichloromethane, ≥99.9%,
66742 Dichloromethane, Selectophore, ≥99.5%,
1.06049 Dichloromethane, EMPROVE® ESSENTIAL Ph Eur,BP,NF, EMPROVE® ESSENTIAL Ph Eur,BP,NF
1.06050 Dichloromethane, for analysis EMSURE® ACS,ISO,Reag. Ph Eur, for analysis EMSURE® ACS,ISO,Reag. Ph Eur
DX0835 Dichloromethane, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS,
1.06054 Dichloromethane, for gas chromatography ECD and FID SupraSolv®, for gas chromatography ECD and FID SupraSolv®
1.06044 Dichloromethane, for liquid chromatography LiChrosolv®, for liquid chromatography LiChrosolv®
DX0831 Dichloromethane, OmniSolv®, OmniSolv®
DX0830 Dichloromethane, Biosynthesis OmniSolv®, Biosynthesis OmniSolv®
DX0837 Dichloromethane, OmniSolv® HR-GC, OmniSolv® HR-GC
1.00668 Dichloromethane, for gas chromatography MS SupraSolv®, for gas chromatography MS SupraSolv®
DX0838 Dichloromethane, HPLC, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs,
1.06048 Dichloromethane, for spectroscopy Uvasol®, for spectroscopy Uvasol®
1.06454 Dichloromethane, for organic trace analysis UniSolv®, for organic trace analysis UniSolv, hypergrade for organic trace analysis SupraSolv®
1.07020 Dichloromethane, for analysis EMPARTA® ACS, for analysis EMPARTA® ACS
8.22271 Dichloromethane, EMPLURA®, EMPLURA®
L090000 Dichloromethane,
L090001 Dichloromethane,
91301 Dichloromethane, Selectophore, ≥99.5% (GC), inhibitor-free,
900633 Dichloromethane, anhydrous, contains 40-150 ppm amylene as stabilizer, ZerO2®, ≥99.8%,
DX0834 Dichloromethane, anhydrous, septum-sealed bottle DriSolv®, anhydrous, septum-sealed bottle DriSolv