• USA Home
  • Search Results

  • dichloromethyloctadecylsilane36751515775511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Dichloro-methyl-octadecylsilane

1 Products
Synonym:   Methyl-octadecyl-dichlorosilane, Octadecyl-methyl-dichlorosilane, Silane ODM
Image of Dichloro-<WBR>methyl-<WBR>octadecylsilane
CAS Number:
Empirical Formula (Hill Notation):
 C19H40Cl2Si
Molecular Weight:
 367.51
 Beilstein/REAXYS Number:
 1774816
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More