• USA Home
  • Search Results

  • dichlorophene269129723411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Dichlorophene

1 Products
Synonym:   Bis(5-chloro-2-hydroxyphenyl)methane, Dichlorophen, Dichlorophene
Image of Dichlorophene
CAS Number:
Linear Formula:
 CH2[C6H3(Cl)OH]2
Molecular Weight:
 269.12
 Beilstein/REAXYS Number:
 1884514
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More