• USA Home
  • Search Results

  • dichlorvos220986273711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Dichlorvos

2 Products
Synonym:   2,2-Dichlorovinyl dimethyl phosphate, DDVP
Image of Dichlorvos
CAS Number:
Empirical Formula (Hill Notation):
 C4H7Cl2O4P
Molecular Weight:
 220.98
 Beilstein/REAXYS Number:
 1709141
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
Show More