• USA Home
  • Search Results

  • diclofenacsodiumsalt318131530779611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Diclofenac sodium salt

4 Products
Synonym:   2-[(2,6-Dichlorophenyl)amino]benzeneacetic acid sodium salt
Image of Diclofenac sodium salt
CAS Number:
Empirical Formula (Hill Notation):
 C14H10Cl2NNaO2
Molecular Weight:
 318.13
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Anal. Standard Form
  Shelf Life
  Assay
  Physical Form
  Special Grade
Show More