• USA Home
  • Search Results

  • dicyclohexylphthalate330428461711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Dicyclohexyl phthalate

3 Products
Image of Dicyclohexyl phthalate
CAS Number:
Linear Formula:
 C6H4-1,2-(CO2C6H11)2
Molecular Weight:
 330.42
 Beilstein/REAXYS Number:
 1889288
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Application(s)
Show More