• USA Home
  • Search Results

  • dicycloverinehydrochloride345956792511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Dicycloverine hydrochloride

2 Products
Synonym:   2-(Diethylamino)ethyl 1-cyclohexylcyclohexane-1-carboxylate hydrochloride, Dicyclomine hydrochloride, [1,1-Bicyclohexyl]-1-carboxylic acid 2-(diethylamino)ethyl ester hydrochloride
Image of Dicycloverine hydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C19H35NO2 · HCl
Molecular Weight:
 345.95
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Manufacturer designation
  Special Grade
Show More