• USA Home
  • Search Results

  • didecyldimethylammoniumchloride36208717351511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Didecyldimethylammonium chloride

1 Products
Synonym:   N-Decyl-N,N-dimethyl-1-decanaminium chloride, DDAC
Image of Didecyldimethylammonium chloride
CAS Number:
Empirical Formula (Hill Notation):
 C22H48ClN
Molecular Weight:
 362.08
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More