• USA Home
  • Search Results

  • didecylphthalate446668477511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Didecyl phthalate

1 Products
Image of Didecyl phthalate
CAS Number:
Linear Formula:
 C6H4[COO(CH2)9CH3]2
Molecular Weight:
 446.66
 Beilstein/REAXYS Number:
 1893077
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More