• USA Home
  • Search Results

  • didymin594561425947311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Didymin

2 Products
Synonym:   (S)-5,7-Dihydroxy-4′-methoxyflavanone-7-β-rutinoside, Isosakuranetin 7-O-rutinoside, Neoponcirin
Image of Didymin
CAS Number:
Empirical Formula (Hill Notation):
 C28H34O14
Molecular Weight:
 594.56
 Beilstein/REAXYS Number:
 1361217
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Application(s)
Show More