• USA Home
  • Search Results

  • diethanolamine1051411142211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Diethanolamine

10 Products
Synonym:   2,2′-Iminodiethanol, Bis(2-hydroxyethyl)amine
Image of Diethanolamine
CAS Number:
Linear Formula:
 HN(CH2CH2OH)2
Molecular Weight:
 105.14
 Beilstein/REAXYS Number:
 605315
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Restriction
  Anal. Standard Form
  UV-Absorption Values
  Solubility
  Product Line
  Viscosity
  Vapor pressure
  Transition temperature
  Melting Point
  Explosion limit
  Density
  Autoignition temp
  Application(s)
  Potency
  pH Value
  Special Grade
  Assay
Show More