• USA Home
  • Search Results

  • diethoxymethylphenylsilane2103477556411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Diethoxy(methyl)phenylsilane

1 Products
Synonym:   Methyl-phenyl-diethoxysilane
Image of Diethoxy(methyl)<WBR>phenylsilane
CAS Number:
Linear Formula:
 C6H5Si(OC2H5)2CH3
Molecular Weight:
 210.34
 Beilstein/REAXYS Number:
 2938396
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More