• USA Home
  • Search Results

  • diethylamine731410989711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Diethylamine

6 Products
Image of Diethylamine
CAS Number:
Linear Formula:
 (C2H5)2NH
Molecular Weight:
 73.14
 Beilstein/REAXYS Number:
 605268
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Restriction
  Refractive Index
  pH Value
  Vapor pressure
  Transition temperature
  Solubility
  Purity Grade
  Melting Point
  Explosion limit
  Density
  Autoignition temp
  Special Grade
  Potency
Show More