• USA Home
  • Search Results

  • diethylaminosulfurtrifluoride161193807809011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(Diethylamino)sulfur trifluoride

1 Products
Synonym:   DAST, Diethylaminosulfur trifluoride
Image of (Diethylamino)sulfur trifluoride
CAS Number:
Linear Formula:
 (C2H5)2NSF3
Molecular Weight:
 161.19
 Beilstein/REAXYS Number:
 1849066
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More