• USA Home
  • Search Results

  • diethylbenzene134222534017411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Diethylbenzene

1 Products
Image of Diethylbenzene
CAS Number:
Linear Formula:
 C6H4(C2H5)2
Molecular Weight:
 134.22
 Beilstein Registry Number:
 1903396
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
D91055 Diethylbenzene, 95%,