• USA Home
  • Search Results

  • diethylchloromethylphosphonate18657316763311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Diethyl chloromethylphosphonate

1 Products
Synonym:   Chloromethylphosphonic acid diethyl ester, NSC 67753
Image of Diethyl chloromethylphosphonate
CAS Number:
Linear Formula:
 ClCH2PO(OC2H5)2
Molecular Weight:
 186.57
 Beilstein/REAXYS Number:
 1363361
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More