• USA Home
  • Search Results

  • diethyleneglycoldiethylether1622311236711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Diethylene glycol diethyl ether

3 Products
Synonym:   2-Ethoxyethyl ether, Bis(2-ethoxyethyl) ether, Diethyldiglycol
Image of Diethylene glycol diethyl ether
CAS Number:
Linear Formula:
 (C2H5OCH2CH2)2O
Molecular Weight:
 162.23
 Beilstein/REAXYS Number:
 1699259
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Potency
  Special Grade
  Explosion limit
  Physical Form
  UV-Absorption Values
  Melting Point
Show More