• USA Home
  • Search Results

  • diethylenetriaminepentaaceticacid393356743611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Diethylenetriaminepentaacetic acid

4 Products
Synonym:   (Carboxymethylimino)bis(ethylenenitrilo)tetraacetic acid, N,N-Bis(2-[bis(carboxymethyl)amino]ethyl)glycine, DETAPAC, DTPA, Penta(carboxymethyl)diethylenetriamine, Pentetic acid
Image of Diethylenetriaminepentaacetic acid
CAS Number:
Linear Formula:
 [(HOOCCH2)2NCH2CH2]2NCH2COOH
Molecular Weight:
 393.35
 Beilstein/REAXYS Number:
 1810219
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More