• USA Home
  • Search Results

  • diethylisocyanomethylphosphonate177144100394511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Diethyl isocyanomethylphosphonate

2 Products
Synonym:   Isocyanomethylphosphonic acid diethyl ester
Image of Diethyl isocyanomethylphosphonate
CAS Number:
Linear Formula:
 CNCH2P(O)(OC2H5)2
Molecular Weight:
 177.14
 Beilstein Registry Number:
 4674920
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More