• USA Home
  • Search Results

  • diethylltartrate206198791211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Diethyl L-tartrate

1 Products
Synonym:   (+)-Diethyl L-tartrate, L-(+)-Tartaric acid diethyl ester
Image of Diethyl <SC>L</SC>-tartrate
CAS Number:
Linear Formula:
 [-CH(OH)CO2C2H5]2
Molecular Weight:
 206.19
 Beilstein/REAXYS Number:
 1727145
 Council of Europe no.:
 440
 EC Number:
 FEMA Number:
 2378
 Flavis number:
 9.446
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
Show More