• USA Home
  • Search Results

  • diethylmaleate1721814105911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Diethyl maleate

3 Products
Synonym:   Maleic acid diethyl ester
Image of Diethyl maleate
CAS Number:
Linear Formula:
 C2H5OCOCH=CHCOOC2H5
Molecular Weight:
 172.18
 Beilstein/REAXYS Number:
 1100825
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Agency/Method
  Special Grade
Show More