• USA Home
  • Search Results

  • diethyloxalate146149592111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Diethyl oxalate

4 Products
Image of Diethyl oxalate
CAS Number:
Linear Formula:
 C2H5OCOCOOC2H5
Molecular Weight:
 146.14
 Beilstein/REAXYS Number:
 606350
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Vapor pressure
  Transition temperature
  Potency
  Melting Point
  Explosion limit
  Density
  Autoignition temp
  Solubility
Show More