• USA Home
  • Search Results

  • diethylphthalate222248466211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Diethyl phthalate

6 Products
Image of Diethyl phthalate
CAS Number:
Linear Formula:
 C6H4-1,2-(CO2C2H5)2
Molecular Weight:
 222.24
 Beilstein/REAXYS Number:
 1912500
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Product Line
  Manufacturer designation
  Anal. Standard Form
  Application(s)
  Assay
  Physical Form
Show More