• USA Home
  • Search Results

  • diethylpyrocarbonate16214160947811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Diethyl pyrocarbonate

3 Products
Synonym:   DEP, DEPC, Diethyl dicarbonate, Diethyl oxydiformate, Ethoxyformic acid anhydride
Image of Diethyl pyrocarbonate
CAS Number:
Linear Formula:
 O(COOC2H5)2
Molecular Weight:
 162.14
 Beilstein/REAXYS Number:
 637031
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Product Information Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Ice Shipping
Show More