• USA Home
  • Search Results

  • diethylstilbestrol268355653111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Diethylstilbestrol

5 Products
Synonym:   (E)-3,4-Bis(4-hydroxyphenyl)-3-hexene, DES, Stilbestrol
Image of Diethylstilbestrol
CAS Number:
Empirical Formula (Hill Notation):
 C18H20O2
Molecular Weight:
 268.35
 Beilstein/REAXYS Number:
 2056095
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Anal. Standard Form
  Physical Form
  Application(s)
  Manufacturer designation
Show More