• USA Home
  • Search Results

  • digitoxin764947163611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Digitoxin

5 Products
Synonym:   5β,20(22)-Cardenolide-3β,14-diol-3-(O-2,6-dideoxy-β-D-ribo-hexopyranosyl-[1→4]-O-2,6-dideoxy-β-D-ribo-hexopyranosyl-[1→4]-2,6-dideoxy-β-D-ribo-hexopyranosyl)oxy, Digitoxoside, Lanatoxin
Image of Digitoxin
CAS Number:
Empirical Formula (Hill Notation):
 C41H64O13
Molecular Weight:
 764.94
 Beilstein/REAXYS Number:
 76678
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Manufacturer designation
  Anal. Standard Form
  Assay
  Physical Form
  Special Grade
Show More