• USA Home
  • Search Results

  • digoxin780942083075511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Digoxin

6 Products
Synonym:   12β-Hydroxydigitoxin
Image of Digoxin
CAS Number:
Empirical Formula (Hill Notation):
 C41H64O14
Molecular Weight:
 780.94
 Beilstein Registry Number:
 77011
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Titertest
  Package quantity
  Anal. Standard Form
  Physical Form
Show More