• USA Home
  • Search Results

  • dihexadecylphosphate54685219763911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Dihexadecyl phosphate

1 Products
Synonym:   DHP, Dicetyl phosphate
Image of Dihexadecyl phosphate
CAS Number:
Linear Formula:
 [CH3(CH2)15O]2P(O)OH
Molecular Weight:
 546.85
 Beilstein/REAXYS Number:
 1717117
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
D2631 Dihexadecyl phosphate,