• USA Home
  • Search Results

  • dihydrokaempferol2882548020611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Dihydrokaempferol

2 Products
Synonym:   (+)-Aromadendrol, (+)-Dihydrokaempferol, (2R,3R)-3,4′,5,7-Tetrahydroxyflavanone, (2R,3R)-3,5,7-Trihydroxy-2-(4-hydroxyphenyl)-2,3-dihydro-4H-chromen-4-one, Aromadendrin, Katuranin
Image of Dihydrokaempferol
CAS Number:
Empirical Formula (Hill Notation):
 C15H12O6
Molecular Weight:
 288.25
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Application(s)
Show More