• USA Home
  • Search Results

  • dihydromyricetin320252720012011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Dihydromyricetin

2 Products
Synonym:   (2R,3R)-3,5,7-Trihydroxy-2-(3,4,5-trihydroxyphenyl)-2,3-dihydrochromen-4-one, 3,3′,4′,5,5′,7-Hexahydroxyflavanone, Ampelopsin, Ampeloptin, DHM
Image of Dihydromyricetin
CAS Number:
Empirical Formula (Hill Notation):
 C15H12O8
Molecular Weight:
 320.25
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More