• USA Home
  • Search Results

  • dihydrostreptomycinsesquisulfate73071549027711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Dihydrostreptomycin sesquisulfate

2 Products
Image of Dihydrostreptomycin sesquisulfate
CAS Number:
Linear Formula:
 C21H41N7O12 · 3/2H2SO4
Molecular Weight:
 730.71
 Beilstein/REAXYS Number:
 3894221
 EC Number:
 MDL number:
 NACRES:
 NA.85
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Activity Spectrum
  Mode of Action
  Physical Form
  Color
Show More