• USA Home
  • Search Results

  • diisobutyladipate2583514104811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Diisobutyl adipate

2 Products
Synonym:   Hexanedioic acid 1,6-bis(2-methylpropyl) ester, NSC 6343
Image of Diisobutyl adipate
CAS Number:
Linear Formula:
 [-CH2CH2CO2CH2CH(CH3)2]2
Molecular Weight:
 258.35
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More