• USA Home
  • Search Results

  • diisobutylaluminumhydridesolution14222119115711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Diisobutylaluminum hydride solution

7 Products
Synonym:   DIBAL-H, DIBAL
Image of Diisobutylaluminum hydride solution
CAS Number:
Linear Formula:
 [(CH3)2CHCH2]2AlH
Molecular Weight:
 142.22
 Beilstein/REAXYS Number:
 4123663
 MDL number:
 PubChem Substance ID:
 eCl@ss:
 38120609
Popular Documents:  Bulletin (PDF)  |  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Density
Show More