• USA Home
  • Search Results

  • diisobutylphthalate278348469511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Diisobutyl phthalate

2 Products
Synonym:   DIBP
Image of Diisobutyl phthalate
CAS Number:
Linear Formula:
 C6H4-1,2-[CO2CH2CH(CH3)2]2
Molecular Weight:
 278.34
 Beilstein/REAXYS Number:
 2054802
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More