• USA Home
  • Search Results

  • diisopropylazodicarboxylate20221244683511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Diisopropyl azodicarboxylate

1 Products
Synonym:   DIAD, Diisopropyl azodiformate
Image of Diisopropyl azodicarboxylate
CAS Number:
Linear Formula:
 (CH3)2CHOOCN=NCOOCH(CH3)2
Molecular Weight:
 202.21
 Beilstein Registry Number:
 1912326
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More