• USA Home
  • Search Results

  • dillapiole2222448431111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Dillapiole

1 Products
Synonym:   6-Allyl-4,5-dimethoxy-1,3-benzodioxole
Image of Dillapiole
CAS Number:
Empirical Formula (Hill Notation):
 C12H14O4
Molecular Weight:
 222.24
 Beilstein/REAXYS Number:
 196037
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More