• USA Home
  • Search Results

  • diltiazemhydrochloride450983328622511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Diltiazem hydrochloride

3 Products
Synonym:   (+)-cis-Diltiazem hydrochloride, (2S,3S)-(+)-cis-3-Acetoxy-5-(2-dimethylaminoethyl)-2,3-dihydro-2-(4-methoxyphenyl)-1,5-benzothiazepin-4(5H)-one hydrochloride, CRD-401
Image of Diltiazem hydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C22H26N2O4S · HCl
Molecular Weight:
 450.98
 Beilstein/REAXYS Number:
 4228706
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Manufacturer designation
  Anal. Standard Form
  Physical Form
  Special Grade
Show More