• USA Home
  • Search Results

  • dimethoxymethyloctylsilane218418571215811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Dimethoxy(methyl)octylsilane

1 Products
Synonym:   Methyl-octyldimethoxysilane, Octylmethyldimethoxysilane
Image of Dimethoxy(methyl)<WBR>octylsilane
CAS Number:
Empirical Formula (Hill Notation):
 C11H26O2Si
Molecular Weight:
 218.41
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More