• USA Home
  • Search Results

  • dimethyl26naphthalenedicarboxylate2442484065311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Dimethyl 2,6-naphthalenedicarboxylate

1 Products
Image of Dimethyl 2,6-<WBR>naphthalenedicarboxylate
CAS Number:
Linear Formula:
 C10H6(CO2CH3)2
Molecular Weight:
 244.24
 Beilstein/REAXYS Number:
 987259
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More