• USA Home
  • Search Results

  • dimethyl3hydroxyglutarate17617725055711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Dimethyl 3-hydroxyglutarate

1 Products
Image of Dimethyl 3-hydroxyglutarate
CAS Number:
Linear Formula:
 CH3OCOCH2CH(OH)CH2COOCH3
Molecular Weight:
 176.17
 Beilstein/REAXYS Number:
 1776666
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More