• USA Home
  • Search Results

  • dimethyl4nitrophthalate2391861022011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Dimethyl 4-nitrophthalate

1 Products
Image of Dimethyl 4-nitrophthalate
CAS Number:
Linear Formula:
 O2NC6H3-1,2-(CO2CH3)2
Molecular Weight:
 239.18
 Beilstein/REAXYS Number:
 1886234
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More