• USA Home
  • Search Results

  • dimethylacetylenedicarboxylate1421176242511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Dimethyl acetylenedicarboxylate

1 Products
Synonym:   Dimethyl 2-butynedioate
Image of Dimethyl acetylenedicarboxylate
CAS Number:
Linear Formula:
 CH3OCOC≡CCO2CH3
Molecular Weight:
 142.11
 Beilstein/REAXYS Number:
 607063
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More