• USA Home
  • Search Results

  • dimethylamine450812440311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Dimethylamine

3 Products
Image of Dimethylamine
CAS Number:
Linear Formula:
 (CH3)2NH
Molecular Weight:
 45.08
 Beilstein/REAXYS Number:
 605257
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  pH Value
  Vapor pressure
  Transition temperature
  Purity Grade
  Potency
  Melting Point
  Physical Form
Show More