• USA Home
  • Search Results

  • dimethylaminomalonatehydrochloride183591611580311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Dimethyl aminomalonate hydrochloride

1 Products
Synonym:   Aminomalonic acid dimethyl ester hydrochloride
Image of Dimethyl aminomalonate hydrochloride
CAS Number:
Linear Formula:
 NH2CH(COOCH3)2 · HCl
Molecular Weight:
 183.59
 Beilstein/REAXYS Number:
 3696467
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More