• USA Home
  • Search Results

  • dimethyldiselenide18799710131711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Dimethyl diselenide

1 Products
Synonym:   Methyl diselenide
Image of Dimethyl diselenide
CAS Number:
Linear Formula:
 (CH3)2Se2
Molecular Weight:
 187.99
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
328502 Dimethyl diselenide, 96%,