• USA Home
  • Search Results

  • dimethyldisulfide942062492011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Dimethyl disulfide

6 Products
Synonym:   DMDS, Methyl disulfide
Image of Dimethyl disulfide
CAS Number:
Linear Formula:
 CH3SSCH3
Molecular Weight:
 94.20
 Beilstein/REAXYS Number:
 1730824
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Refractive Index
  Document Links
  Allergen
  Organoleptic property
  Application(s)
  Biological Source
  Agency/Method
  Special Grade
Show More