• USA Home
  • Search Results

  • dimethylfumarate1441362449711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Dimethyl fumarate

4 Products
Image of Dimethyl fumarate
CAS Number:
Linear Formula:
 CH3OCOCH=CHCOOCH3
Molecular Weight:
 144.13
 Beilstein/REAXYS Number:
 774590
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Application(s)
  Potency
  Assay
Show More