• USA Home
  • Search Results

  • dimethylisophthalate19418145993411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Dimethyl isophthalate

2 Products
Image of Dimethyl isophthalate
CAS Number:
Linear Formula:
 C6H4-1,3-(CO2CH3)2
Molecular Weight:
 194.18
 Beilstein/REAXYS Number:
 1912251
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Anal. Standard Form
  Shelf Life
  Physical Form
  Purity Grade
  Special Grade
  Titertest
Show More