• USA Home
  • Search Results

  • dimethylsebacate2303010679611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Dimethyl sebacate

2 Products
Synonym:   Decanedioic acid 1,10-dimethyl ester, Dimethyl decanedioate, NSC 9415, Sebacic acid dimethyl ester
Image of Dimethyl sebacate
CAS Number:
Linear Formula:
 CH3OCO(CH2)8COOCH3
Molecular Weight:
 230.30
 Beilstein/REAXYS Number:
 1785523
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More