• USA Home
  • Search Results

  • dimethylsulfoxide78136768511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Dimethyl sulfoxide

27 Products
Synonym:   DMSO
Image of Dimethyl sulfoxide
CAS Number:
Linear Formula:
 (CH3)2SO
Molecular Weight:
 78.13
 Beilstein/REAXYS Number:
 506008
 EC Number:
 MDL number:
 NACRES:
 NA.75
 PubChem Substance ID:
Popular Documents:  Product Information Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Anal. Standard Form
  UV-Absorption Condition
  Sterilization
  Package quantity
  Manufacturer designation
  Density
  Quality clarifier
  Melting Point
  Shelf Life
  Restriction
  Refractive Index
  Agency/Method
  Suitability
  Purity Grade
  Viscosity
  UV-Absorption Values
  Transmittance
  Transition temperature
  Product Line
  Potency
  Color
  Special Grade
  Solubility
  Application(s)
  Physical Form
Show More

Product #
Description
D2650 Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%,
D8418 Dimethyl sulfoxide, for molecular biology,
276855 Dimethyl sulfoxide, anhydrous, ≥99.9%,
D2438 Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma,
472301 Dimethyl sulfoxide, ACS reagent, ≥99.9%,
D4540 Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture,
34869 Dimethyl sulfoxide, suitable for HPLC, ≥99.7%,
D5879 Dimethyl sulfoxide, ReagentPlus®, ≥99.5%,
D1435 Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications,
1.01900 Dimethyl sulfoxide, for headspace gas chromatography SupraSolv®,
41640 Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC),
41639 Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC),
D9170 Dimethyl sulfoxide, PCR Reagent,
1.02952 Dimethyl sulfoxide, for analysis EMSURE® ACS,
900645 Dimethyl sulfoxide, anhydrous, ZerO2®, ≥99.9%,
34943 Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water,
01934 Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis),
1.02950 Dimethyl sulfoxide, for spectroscopy Uvasol®,
1.37117 Dimethyl sulfoxide, EMPROVE® EXPERT Ph Eur,USP,
94563 Dimethyl sulfoxide, analytical standard,
5.43900 Dimethyl sulfoxide, for liquid chromatography LiChrosolv®,
1.09678 Dimethyl sulfoxide, for gas chromatography,
1.02931 Dimethyl sulfoxide, for DNA and peptide synthesis (max. 0.025% H₂O),
1.16743 Dimethyl sulfoxide, EMPLURA®,
1211006 Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard,
D2050000 Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard,
RTC000103 Dimethyl Sulfoxide, Pharmaceutical Secondary Standard; Certified Reference Material,